Learn More
Med Vet International Surgical Drape 3M Steri-Drape Isolation Drape 20 W X 20 L inch Sterile, 1003, Each
Catalog No. 502830845
Encompass_Preferred
Film is durable and resistant to fluid strike-through to keep contents secure, yet is soft enough to be gentle on organs and tissues Translucent plastic film allows for visualization of contents 20 x 20 inches
Catalog No. 50-283-0845 Supplier Med Vet International Supplier No. 142526EA
Film is durable and resistant to fluid strike-through to keep contents secure, yet is soft enough to be gentle on organs and tissues Translucent plastic film allows for visualization of contents 20 x 20 inches