Learn More
Med Vet International Surgical Drape 3M Steri-Drape Small Drape With Aperture 22 W X 25 L inch Sterile, 1092, 25/Box
Catalog No. 502829480
Encompass_Preferred
The sterile surgical drapes with polyethylene film create an impenetrable barrier that resists fluid leaks They minimize the risk of infection by reducing fluid exposure to the skin The aperture with adhesive conforms to the body producing a reliable hold They prevent patient infection at the wound site and cross-contamination The disposable drape sheets have a matte finish to reduce glare
Catalog No. 50-282-9480 Supplier Med Vet International Supplier No. 5727BX
Add to Cart
Add to Cart
The sterile surgical drapes with polyethylene film create an impenetrable barrier that resists fluid leaks They minimize the risk of infection by reducing fluid exposure to the skin The aperture with adhesive conforms to the body producing a reliable hold They prevent patient infection at the wound site and cross-contamination The disposable drape sheets have a matte finish to reduce glare