Learn More
Med Vet International Disposable Surgical Veterinary Drape SMS 40in X 100 Yards, Blue/Gray, each
Catalog No. 502522171
Encompass_Preferred
This high quality surgical drape is made of SMS non-woven material. Consists of two blue/gray outer layers of SMS and a middle layer of spunbound polypropylene. Our disposable vet drape can surround the surgical area with barrier proof material during veterinary surgery. Can also be used as a drape to control and contain fluid, as fenestrated material or as a sterilization wrap. Fabric is low lint, helping to reduce wound contamination. Microfibers filter out dry particles and many water-based liquids. First-rate option for a variety of innovative surgical draping solutions and also for general medical applications. Cloth-like feel Autoclavable.
Catalog No. 50-252-2171 Supplier Med Vet International Supplier No. SMS40
This high quality surgical drape is made of SMS non-woven material. Consists of two blue/gray outer layers of SMS and a middle layer of spunbound polypropylene. Our disposable vet drape can surround the surgical area with barrier proof material during veterinary surgery. Can also be used as a drape to control and contain fluid, as fenestrated material or as a sterilization wrap. Fabric is low lint, helping to reduce wound contamination. Microfibers filter out dry particles and many water-based liquids. First-rate option for a variety of innovative surgical draping solutions and also for general medical applications. Cloth-like feel Autoclavable.