Learn More
Med Vet International General Purpose Drape 3M Steri-Drape Drape Sheet / Back Table Cover 44 W X 59 L inch Sterile, 9072, 80/Case
Catalog No. 502831619
Encompass_Preferred
3M general-purpose drapes for general surgery, orthopedic, neurological, ophthalmic and cardiovascular procedures Strong fabric is resistant to fluid leaks and strikethrough 3M's Steri-Drape sheets promote a safe environment Low-linting design helps reduce the transfer of bacteria This Steri-Drape is 44 inches wide x 59 inches long A 2-inch adhesive strip on the back holds them in place
Catalog No. 50-283-1619 Supplier Med Vet International Supplier No. 449931CS
3M general-purpose drapes for general surgery, orthopedic, neurological, ophthalmic and cardiovascular procedures Strong fabric is resistant to fluid leaks and strikethrough 3M's Steri-Drape sheets promote a safe environment Low-linting design helps reduce the transfer of bacteria This Steri-Drape is 44 inches wide x 59 inches long A 2-inch adhesive strip on the back holds them in place