Learn More
Med Vet International 3M Steri-Drape Instrument Pocket Drape, 1018, 40/Case
Catalog No. 502830456
Encompass_Preferred
2 Compartments Plastic pouch used to organize and secure instruments in the sterile field Adhesive strips on each end of the pouch hold it in place
Catalog No. 50-283-0456 Supplier Med Vet International Supplier No. 94540CS
2 Compartments Plastic pouch used to organize and secure instruments in the sterile field Adhesive strips on each end of the pouch hold it in place