Learn More
Med Vet International General Purpose Drape 3M Steri-Drape Large Towel Drape 17 W X 23 L inch Sterile, 1010, 40/Case
Catalog No. 502831631
Encompass_Preferred
Impervious polyethylene film provides a barrier to liquids, reducing exposure to blood and body fluids Reliable adhesive stays secure throughout the procedure Drape can be removed easily, enhancing patient comfort and minimizing clean-up time Matte finish helps reduce glare from drape surface Adheres to dry skin to help prevent fluids from running under drape and patient, minimizing the risk of body fluid exposure to healthcare personnel and making clean up easier 17 inches wide x 23 inches long
Catalog No. 50-283-1631 Supplier Med Vet International Supplier No. 5714CS
Impervious polyethylene film provides a barrier to liquids, reducing exposure to blood and body fluids Reliable adhesive stays secure throughout the procedure Drape can be removed easily, enhancing patient comfort and minimizing clean-up time Matte finish helps reduce glare from drape surface Adheres to dry skin to help prevent fluids from running under drape and patient, minimizing the risk of body fluid exposure to healthcare personnel and making clean up easier 17 inches wide x 23 inches long