Learn More
Med Vet International Orthopedic Drape 3M Steri-Drape U-Drape 47 W X 51 L inch Sterile, 1015, 40/Case
Catalog No. 502830929
Encompass_Preferred
Impervious plastic film provides a barrier to liquids, reducing exposure to blood and body fluids Reliable adhesive around slot stays secure throughout the procedure Polyethylene film is transparent and conformable for easy positioning and draping, helping to save time Matte finish reduces glare from drape surface during procedure U drape has a U slot aperture with adhesive 47 inches wide x 51 inches long
Catalog No. 50-283-0929 Supplier Med Vet International Supplier No. 5716CS
Impervious plastic film provides a barrier to liquids, reducing exposure to blood and body fluids Reliable adhesive around slot stays secure throughout the procedure Polyethylene film is transparent and conformable for easy positioning and draping, helping to save time Matte finish reduces glare from drape surface during procedure U drape has a U slot aperture with adhesive 47 inches wide x 51 inches long