Learn More
Med Vet International Orthopedic Drape 3M Steri-Drape U-Pouch 35 W X 30 L inch Sterile, 1067, Each
Catalog No. 502829472
Encompass_Preferred
Matte finish reduces glare from drape surface during procedure U-pouch is made of an impervious polyethylene film to prevent leakage of fluid from the pouch Reliable adhesive around slot stays secure throughout the procedure 35 inches wide x 30 inches long
Catalog No. 50-282-9472 Supplier Med Vet International Supplier No. 177162EA
Matte finish reduces glare from drape surface during procedure U-pouch is made of an impervious polyethylene film to prevent leakage of fluid from the pouch Reliable adhesive around slot stays secure throughout the procedure 35 inches wide x 30 inches long