Learn More
Med Vet International General Purpose Drape 3M Steri-Drape Utility Sheet 22 W X 25 L inch Sterile, 1089, 2/Pack
Catalog No. 502831495
Encompass_Preferred
These 3M general-purpose drapes are super absorbent to prevent fluid leaks They have an adhesive backing that produces a strong bond for reliable use The 3M Steri-Drape sheets are 22 inches wide x 25 inches long The modular design and adhesive backing let them attach to other drapes They're made of a low-lint material that helps to reduce contamination at the site The sterile utility sheets are suitable for major and minor surgeries
Catalog No. 50-283-1495 Supplier Med Vet International Supplier No. 362568PK
These 3M general-purpose drapes are super absorbent to prevent fluid leaks They have an adhesive backing that produces a strong bond for reliable use The 3M Steri-Drape sheets are 22 inches wide x 25 inches long The modular design and adhesive backing let them attach to other drapes They're made of a low-lint material that helps to reduce contamination at the site The sterile utility sheets are suitable for major and minor surgeries