Learn More
Med Vet International Sontara Surgical Drape, 59in x 100 Yards, Disposable, Each
Catalog No. 502091808
Encompass_Preferred
The original Sontara surgical drape comes on a 59in. x 100-yard roll and has a rich blue color. This disposable surgical drape is made without the use of binders, chemicals, or adhesive, minimizing the potential for contamination. Sontara surgical drapes are opaque which helps to minimize glare, are strong and tear-resistant, and provide optimal liquid management. Key Features of this surgical drape are: Low Linting Provide optimal liquid management Soft High strength
Catalog No. 50-209-1808 Supplier Med Vet International Supplier No. MVD60
The original Sontara surgical drape comes on a 59in. x 100-yard roll and has a rich blue color. This disposable surgical drape is made without the use of binders, chemicals, or adhesive, minimizing the potential for contamination. Sontara surgical drapes are opaque which helps to minimize glare, are strong and tear-resistant, and provide optimal liquid management. Key Features of this surgical drape are: Low Linting Provide optimal liquid management Soft High strength