Learn More
Med Vet International Sontara Surgical Drape, 39in x 100 Yards, Disposable, Each
Catalog No. 502091809
Encompass_Preferred
The original Sontara surgical drape comes on a 39in. x 100-yard roll and has a rich blue color. This disposable surgical drape is made without the use of binders, chemicals, or adhesive, minimizing the potential for contamination. Sontara surgical drapes are opaque which helps to minimize glare, are strong and tear-resistant, and provide optimal liquid management. Key Features of this surgical drape are: Low Linting Provide optimal liquid management Soft High strength
Catalog No. 50-209-1809 Supplier Med Vet International Supplier No. MVD40
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
The original Sontara surgical drape comes on a 39in. x 100-yard roll and has a rich blue color. This disposable surgical drape is made without the use of binders, chemicals, or adhesive, minimizing the potential for contamination. Sontara surgical drapes are opaque which helps to minimize glare, are strong and tear-resistant, and provide optimal liquid management. Key Features of this surgical drape are: Low Linting Provide optimal liquid management Soft High strength