Learn More
Med Vet International Fenstrated Drape, 30"X30", 2"X1" Rectangle, Each
Catalog No. 502091813
Encompass_Preferred
Cloth fenestrated drapes made from high quality surgical sheeting with built-in moisture barrier. Drapes are colorfast, washable, autoclavable and come in a choice of sizes.
Catalog No. 50-209-1813 Supplier Med Vet International Supplier No. 30302
Cloth fenestrated drapes made from high quality surgical sheeting with built-in moisture barrier. Drapes are colorfast, washable, autoclavable and come in a choice of sizes.