Learn More
Med Vet International Fenestrated Drape, 40"X40", 10"X1.25", Rectangle, Each
Catalog No. 502091817
Encompass_Preferred
Cloth fenestrated drapes made from high quality surgical sheeting with built-in moisture barrier. Drapes are colorfast, washable, and autoclavable.
Catalog No. 50-209-1817 Supplier Med Vet International Supplier No. 404010
Add to Cart
Add to Cart
Cloth fenestrated drapes made from high quality surgical sheeting with built-in moisture barrier. Drapes are colorfast, washable, and autoclavable.