Learn More
Med Vet International Drape Medium Tape 40X70 Sterile 50 Ea/Case
Catalog No. 502091828
Encompass_Preferred
Medline's surgical drapes have received an upgrade! Competitive with the new industry standard, a high-quality SMS material provides exceptional barrier protection (double the hydrostatic pressure of Eclipse fabric) without sacrificing comfort or softness. Our SMS fabric binds three layers of fabric--a densely-packed meltblown layer sandwiched between a strong spunbond outer layers--to provide incredible fluid repellency. Progressive Reicofil technology offers customers increased fabric uniformity and drapability, equipping you with the market's latest-generation of SMS. You really can't get more from a surgical drape. Medium Drape w/Tape 70"x40".
Catalog No. 50-209-1828 Supplier Med Vet International Supplier No. DYNJP2412T
Medline's surgical drapes have received an upgrade! Competitive with the new industry standard, a high-quality SMS material provides exceptional barrier protection (double the hydrostatic pressure of Eclipse fabric) without sacrificing comfort or softness. Our SMS fabric binds three layers of fabric--a densely-packed meltblown layer sandwiched between a strong spunbond outer layers--to provide incredible fluid repellency. Progressive Reicofil technology offers customers increased fabric uniformity and drapability, equipping you with the market's latest-generation of SMS. You really can't get more from a surgical drape. Medium Drape w/Tape 70"x40".