Learn More
Cole-Parmer PolarSafe Upright Freezer Drawer Rack for Standard 2" Boxes, 4 x 6 Array
Catalog No. NC1069046 Shop All Cole-Parmer Products
Encompass_Preferred
Drawers slide out independently for easy access to the boxes. Use with standard 2" cardboard, plastic, or metal boxes. Built-in labelling card holders help identify drawer content. Designed to fit all major brands of ultralow upright freezers. Sturdy corrosion-resistant stainless steel construction for long-term use.
Catalog No. NC1069046 Supplier Cole-Parmer Supplier No. 0439823
Drawers slide out independently for easy access to the boxes. Use with standard 2" cardboard, plastic, or metal boxes. Built-in labelling card holders help identify drawer content. Designed to fit all major brands of ultralow upright freezers. Sturdy corrosion-resistant stainless steel construction for long-term use.