Learn More
Grainger Drawer 22 in L x 16 in W Steel White
Catalog No. 502751490
Encompass_Preferred
The Knaack 14V869 is a 14-inch wide, 8-inch deep, and 69-inch high steel job box with a lockable drawer. It features a continuous piano hinge, gas struts for easy opening, and a hasp for padlocking. The body is made from 20 gauge steel with a powder coat finish. The load capacity is 500 lbs. This job box provides secure storage for tools and equipment on jobsites and in work vehicles. With its durable steel construction and lockable drawer, it helps protect contents from weather, impacts, and theft.
Catalog No. 50-275-1490 Supplier Grainger Supplier No. 14V869
The Knaack 14V869 is a 14-inch wide, 8-inch deep, and 69-inch high steel job box with a lockable drawer. It features a continuous piano hinge, gas struts for easy opening, and a hasp for padlocking. The body is made from 20 gauge steel with a powder coat finish. The load capacity is 500 lbs. This job box provides secure storage for tools and equipment on jobsites and in work vehicles. With its durable steel construction and lockable drawer, it helps protect contents from weather, impacts, and theft.