Learn More
Alkali Scientific Drawer Rack For 15mL and 50mL Centrifuge Tube Boxes, 3x2 configuration, 6 5" Box Capacity
SDP
Catalog No. 5000662552
Encompass_Preferred
This drawer rack is designed for efficient storage and organization of 15 mL and 50 mL centrifuge tube boxes in laboratory refrigerators and freezers. The practical 3×2 configuration securely holds 5–6 standard tube boxes while optimizing drawer space and maintaining easy access to samples. Its sturdy construction supports dependable performance in demanding laboratory environments, making it suitable for clinical, research, and pharmaceutical applications. The rack helps improve sample organization while maximizing storage capacity within refrigeration units. Overall dimensions measure 18 7/16 × 10 1/2 × 6 1/8 inches (478 × 267 × 155 mm), with compartment dimensions of 6 × 5 1/16 × 5 13/16 inches (152.4 × 128.6 × 147.6 mm). It holds six 5 inch boxes arranged in a 3×2 configuration. Locking device DD25 09 is available separately.
Catalog No. 50-006-62552 Supplier Alkali Scientific Supplier No. UFDLB32
This drawer rack is designed for efficient storage and organization of 15 mL and 50 mL centrifuge tube boxes in laboratory refrigerators and freezers. The practical 3×2 configuration securely holds 5–6 standard tube boxes while optimizing drawer space and maintaining easy access to samples. Its sturdy construction supports dependable performance in demanding laboratory environments, making it suitable for clinical, research, and pharmaceutical applications. The rack helps improve sample organization while maximizing storage capacity within refrigeration units. Overall dimensions measure 18 7/16 × 10 1/2 × 6 1/8 inches (478 × 267 × 155 mm), with compartment dimensions of 6 × 5 1/16 × 5 13/16 inches (152.4 × 128.6 × 147.6 mm). It holds six 5 inch boxes arranged in a 3×2 configuration. Locking device DD25 09 is available separately.