Learn More
Alkali Scientific Drawer Rack For 15mL and 50mL Centrifuge Tube Boxes, 4x2 configuration, 8 5" Box Capacity
SDP
Catalog No. 5000662471
Encompass_Preferred
This drawer rack is engineered to provide organized storage for 15 mL and 50 mL centrifuge tube boxes in laboratory refrigerators and freezers. Featuring a spacious 4×2 configuration, the rack accommodates up to 8–10 standard tube boxes while maximizing available drawer capacity and maintaining easy access to stored samples. Built with durable materials, the rack offers dependable long term performance in clinical, research, and pharmaceutical laboratory environments where efficient sample organization is essential. The unit measures 24 1/2 × 10 1/2 × 6 1/8 inches (628 × 267 × 155 mm), and each compartment measures 6 × 5 1/16 × 5 13/16 inches (152.4 × 128.6 × 147.6 mm). It holds eight 5 inch boxes arranged in a 4×2 configuration. Locking device DD25 09 is available separately.
Catalog No. 50-006-62471 Supplier Alkali Scientific Supplier No. UFDLB42
This drawer rack is engineered to provide organized storage for 15 mL and 50 mL centrifuge tube boxes in laboratory refrigerators and freezers. Featuring a spacious 4×2 configuration, the rack accommodates up to 8–10 standard tube boxes while maximizing available drawer capacity and maintaining easy access to stored samples. Built with durable materials, the rack offers dependable long term performance in clinical, research, and pharmaceutical laboratory environments where efficient sample organization is essential. The unit measures 24 1/2 × 10 1/2 × 6 1/8 inches (628 × 267 × 155 mm), and each compartment measures 6 × 5 1/16 × 5 13/16 inches (152.4 × 128.6 × 147.6 mm). It holds eight 5 inch boxes arranged in a 4×2 configuration. Locking device DD25 09 is available separately.