Learn More
Alkali Scientific Drawer Rack For 15mL and 50mL Centrifuge Tube Boxes, 5x2 configuration, 10 5" Box Capacity
SDP
Catalog No. 5000662553
Encompass_Preferred
This drawer rack is designed for high capacity storage and organization of 15 mL and 50 mL centrifuge tube boxes within laboratory refrigerators and freezers. Featuring a 5×2 configuration, the rack accommodates up to 10–12 standard tube boxes while optimizing drawer space and ensuring convenient access to stored samples. Its durable construction supports dependable long term performance in clinical, research, and pharmaceutical laboratory environments where organized, high density storage is essential. The rack measures 30 1/2 × 10 1/2 × 6 1/8 inches (773 × 267 × 155 mm), with compartment dimensions of 6 × 5 1/16 × 5 13/16 inches (152.4 × 128.6 × 147.6 mm). It holds ten 5 inch boxes arranged in a 5×2 configuration. Locking device DD25 09 is available separately.
Catalog No. 50-006-62553 Supplier Alkali Scientific Supplier No. UFDLB52
This drawer rack is designed for high capacity storage and organization of 15 mL and 50 mL centrifuge tube boxes within laboratory refrigerators and freezers. Featuring a 5×2 configuration, the rack accommodates up to 10–12 standard tube boxes while optimizing drawer space and ensuring convenient access to stored samples. Its durable construction supports dependable long term performance in clinical, research, and pharmaceutical laboratory environments where organized, high density storage is essential. The rack measures 30 1/2 × 10 1/2 × 6 1/8 inches (773 × 267 × 155 mm), with compartment dimensions of 6 × 5 1/16 × 5 13/16 inches (152.4 × 128.6 × 147.6 mm). It holds ten 5 inch boxes arranged in a 5×2 configuration. Locking device DD25 09 is available separately.