Learn More
Alkali Scientific Stainless Steel Freezer Racks with Drawers for 2" Boxes, 4 X 4 Configuration, 16 Box Capacity
SDP
Catalog No. 502336018
Encompass_Preferred
The 4 X 4 freezer rack is compatible with standard 2 inch freezer boxes and can hold up to 9 boxes. They are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left-hand doors. These freezer racks have been configured to fit all major ultra-low (-80°) freezers on the market; racks are designed for ultra-low upright freezers. Upright freezer drawer racks offer a more organized system for storage and retrieval of samples. Drawer racks facilitate both access to and storage of samples without having to remove the whole rack from the freezer. Dimensions (D x H x W): 22 5/16 x 9 7/16 x 5 1/2 inches (566 x 240 x 140mm). Compartment Dimensions: 5 3/8 x 2 1/8 x 5 1/4 inches (137.4 x 54.4 x 133mm).
Catalog No. 50-233-6018 Supplier Alkali Scientific Supplier No. UFDR442
The 4 X 4 freezer rack is compatible with standard 2 inch freezer boxes and can hold up to 9 boxes. They are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left-hand doors. These freezer racks have been configured to fit all major ultra-low (-80°) freezers on the market; racks are designed for ultra-low upright freezers. Upright freezer drawer racks offer a more organized system for storage and retrieval of samples. Drawer racks facilitate both access to and storage of samples without having to remove the whole rack from the freezer. Dimensions (D x H x W): 22 5/16 x 9 7/16 x 5 1/2 inches (566 x 240 x 140mm). Compartment Dimensions: 5 3/8 x 2 1/8 x 5 1/4 inches (137.4 x 54.4 x 133mm).