Learn More
CRYSTAL TECHNOLOGY AND INDUSTRIES Rack with Drawer for 50ml Centrifuge Tubes
Catalog No. NC1668830
Encompass_Preferred
Outside Dimensions(LxHxW):16 1/2 x 9 15/16 x 5 1/2 inch in ( 419 x 252 x 140 mm ).This rack has the capacity for 60 tubes (50mL).
Catalog No. NC1668830 Supplier CRYSTAL TECHNOLOGY AND INDUSTRIES Supplier No. UFDT502
Outside Dimensions(LxHxW):16 1/2 x 9 15/16 x 5 1/2 inch in ( 419 x 252 x 140 mm ).This rack has the capacity for 60 tubes (50mL).