Learn More
Alkali Scientific Stainless Steel Freezer Racks with Drawers for 2" Boxes, 3 x 5 Configuration, 15 Box Capacity
SDP
Catalog No. 502388675
Encompass_Preferred
Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left-hand doors. Our freezer racks have been configured to fit all major ultra-low (-80°) freezers on the market; racks are designed for ultra-low upright freezers. Upright Freezer Drawer racks offer a more organized system for storage and retrieval of samples. Drawer racks facilitate both access to and storage of samples without having to remove the whole rack from the freezer. Security lock devices are also available for each rack though ordered separately. Dimensions (D x H x W): 16 3/4 x 11 13/16 x 5 1/2 inches (426 x 300 x 140mm). Compartment Dimensions: 5 3/8 x 2 1/8 x 5 1/4 inches (137.4 x 54.4 x 133mm).
Catalog No. 50-238-8675 Supplier Alkali Scientific Supplier No. UFDR352
Add to Cart
Add to Cart
Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left-hand doors. Our freezer racks have been configured to fit all major ultra-low (-80°) freezers on the market; racks are designed for ultra-low upright freezers. Upright Freezer Drawer racks offer a more organized system for storage and retrieval of samples. Drawer racks facilitate both access to and storage of samples without having to remove the whole rack from the freezer. Security lock devices are also available for each rack though ordered separately. Dimensions (D x H x W): 16 3/4 x 11 13/16 x 5 1/2 inches (426 x 300 x 140mm). Compartment Dimensions: 5 3/8 x 2 1/8 x 5 1/4 inches (137.4 x 54.4 x 133mm).