Learn More
Thomas Scientific Drawer Style Upright Freezer Rack, Holds 18 Matrix Racks. Stainless Steel, 8"H x 21"D x 5 1/2"W.
SDP
Catalog No. 50196042
Encompass_Preferred
For Micronic RoboRack-96 with High Cover Outside Dimensions: 21 1/5 x 10 3/5 x 5 7/16 inch in(538.5 x 268.5 x 138 mm) Internal Dimensions: 3 7/16 x 2 7/16 x 5 1/8 in(88 x 62.4 x 130 mm)
Catalog No. 50-196-042 Supplier Thomas Scientific Supplier No. 1148X02
For Micronic RoboRack-96 with High Cover Outside Dimensions: 21 1/5 x 10 3/5 x 5 7/16 inch in(538.5 x 268.5 x 138 mm) Internal Dimensions: 3 7/16 x 2 7/16 x 5 1/8 in(88 x 62.4 x 130 mm)