Learn More
IMEB, Inc Tek-Select Metal Slide Storage Cabinet Stackable/Lockable for Safety
SDP
Catalog No. NC1154805
Encompass
Tek-Select economical light weight metal microscope slide allow for a secure and safe way of storing microscope slides and paraffin blocks conveniently together or stacked separately. Each metal file system is interlocking and can be stacked 12-15" high. Slides Storage Cabinet Stackable/Lockable for safety. Each unit contains 14 drawers and accommodates approx. 5,600 slides. Paraffin Block Storage Cabinet also available for sale. External dimensions: 12.6" H x 48.5W" x 48"D
Catalog No. NC1154805 Supplier IMEB, Inc Supplier No. SC001
Tek-Select economical light weight metal microscope slide allow for a secure and safe way of storing microscope slides and paraffin blocks conveniently together or stacked separately. Each metal file system is interlocking and can be stacked 12-15" high. Slides Storage Cabinet Stackable/Lockable for safety. Each unit contains 14 drawers and accommodates approx. 5,600 slides. Paraffin Block Storage Cabinet also available for sale. External dimensions: 12.6" H x 48.5W" x 48"D