Learn More
Waters Corp Waters Drawoff Valve Assembly, HPLC, Mobile Phase Priming
Catalog No. 50793471 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Drawoff Valve Assembly is the mechanism responsible for priming the pump heads with mobile phase in the 515 HPLC pump, the 1515 HPLC pump, and the 1525 HPLC pump.
  • Mechanism for priming pump heads with mobile phase.
  • Compatible with 515, 1515, and 1525 HPLC pumps.
Catalog No. 50-793-471 Supplier Waters Corp Supplier No. WAS207085
The Drawoff Valve Assembly is the mechanism responsible for priming the pump heads with mobile phase in the 515 HPLC pump, the 1515 HPLC pump, and the 1525 HPLC pump.
  • Mechanism for priming pump heads with mobile phase.
  • Compatible with 515, 1515, and 1525 HPLC pumps.