Learn More
TARGETMOL CHEMICALS INC DRB 100MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T7789100MG
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DRB is a nucleoside analog that inhibits several carboxyl-terminal domain (CTD) kinases including casein kinase II (IC50 range of 4-10 uM)purity: 99%
Catalog No. 50-257-9122