Learn More
Abcam DRB (Benzimidazole), Casein kinase II inhibitor, 50MG
Catalog No. 502707106
Encompass_Preferred
MW 319.14 g/mol, Purity >98%. Casein kinase II inhibitor (Ki values are 23 and 48 μM for CKII and CKI, respectively). Inhibitor of RNA polymerase II transcription in vitro and in vivo. Shown to inhibit cyclin C/CDK8 (IC₅₀ = 17 μM) and cyclin H/CDK7/p38 (IC₅₀ = 18 μM) signaling.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7106 Supplier Abcam Supplier No. AB12093950MG
MW 319.14 g/mol, Purity >98%. Casein kinase II inhibitor (Ki values are 23 and 48 μM for CKII and CKI, respectively). Inhibitor of RNA polymerase II transcription in vitro and in vivo. Shown to inhibit cyclin C/CDK8 (IC₅₀ = 17 μM) and cyclin H/CDK7/p38 (IC₅₀ = 18 μM) signaling.
The product is subject to the following: Abcam Restricted Use Statement