Learn More
Medchemexpress LLC DRB18 | 2863686-81-9 | 99.6% | C22H23ClN2O2 | 100 MG
SDP
Catalog No. 5000355769
Encompass_Preferred
DRB18 is a potent pan-class GLUT inhibitor that alters energy-related metabolism in A549 cells by changing the abundance of metabolites in glucose-related pathways. It can lead to G1/S phase arrest, increase oxidative stress, and cause necrotic cell death, while also exhibiting anti-tumor activity.
  • Potent pan-class GLUT inhibitor
  • Alters energy-related metabolism in A549 cells
  • Leads to G1/S phase arrest
  • Increases oxidative stress
  • Causes necrotic cell death
  • Exhibits anti-tumor activity
  • Reduces glucose uptake in GLUT1-4-expressed HEK293 cell lines
  • Decreases expression of GLUT1-4 in xenografted tumors
  • Reduces proliferative capacity within xenografted tumors
Catalog No. 50-003-55769 Supplier Medchemexpress LLC Supplier No. HY145963100MG
DRB18 is a potent pan-class GLUT inhibitor that alters energy-related metabolism in A549 cells by changing the abundance of metabolites in glucose-related pathways. It can lead to G1/S phase arrest, increase oxidative stress, and cause necrotic cell death, while also exhibiting anti-tumor activity.
  • Potent pan-class GLUT inhibitor
  • Alters energy-related metabolism in A549 cells
  • Leads to G1/S phase arrest
  • Increases oxidative stress
  • Causes necrotic cell death
  • Exhibits anti-tumor activity
  • Reduces glucose uptake in GLUT1-4-expressed HEK293 cell lines
  • Decreases expression of GLUT1-4 in xenografted tumors
  • Reduces proliferative capacity within xenografted tumors