Learn More
Medchemexpress LLC DRB18 | 2863686-81-9 | 99.6% | C22H23ClN2O2 | 50 MG
SDP
Catalog No. 5000355799
Encompass_Preferred
DRB18 is a potent pan-class GLUT inhibitor that modifies energy-related metabolism in A549 cells by altering metabolite abundance in glucose-related pathways. This can result in G1/S phase arrest, increased oxidative stress, and necrotic cell death. It has demonstrated anti-tumor activity.
  • Potent pan-class GLUT inhibitor
  • Modifies energy-related metabolism in A549 cells
  • Leads to G1/S phase arrest
  • Increases oxidative stress
  • Causes necrotic cell death
  • Demonstrates anti-tumor activity
  • Reduces glucose uptake in HEK293 cell lines
  • Inhibits tumor volume and weight in xenografted tumors
Catalog No. 50-003-55799 Supplier Medchemexpress LLC Supplier No. HY14596350MG
DRB18 is a potent pan-class GLUT inhibitor that modifies energy-related metabolism in A549 cells by altering metabolite abundance in glucose-related pathways. This can result in G1/S phase arrest, increased oxidative stress, and necrotic cell death. It has demonstrated anti-tumor activity.
  • Potent pan-class GLUT inhibitor
  • Modifies energy-related metabolism in A549 cells
  • Leads to G1/S phase arrest
  • Increases oxidative stress
  • Causes necrotic cell death
  • Demonstrates anti-tumor activity
  • Reduces glucose uptake in HEK293 cell lines
  • Inhibits tumor volume and weight in xenografted tumors