Learn More
Selleck Chemical LLC DRB18 5mg
SDP
Supplier:  Selleck Chemical LLC S96365MG
Encompass_Preferred
DRB18 is a pan-GLUT (glucose transporters) inhibitor that inhibits growth and proliferation in several different cancer types with IC50 high nM range. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3481