Learn More
Hach Lange™ DRB200: Digital Reactor Block: 16 mm vial wells, 230 Vac
Catalog No. 501997838 Shop All Hach Company Products
Encompass
Change view
Click to view available options
Size:
Dual block (2 x 15 wells)
Catalog No. Size
50-199-7838 Dual block (2 x 15 wells)
1 options
Catalog No. 50-199-7838 Supplier Hach Lange™ Supplier No. LTV082.52.44001
The simple solution for standard and special digestions.
The simple solution for standard and special digestions. Single and double block options available. Easy-to-use and fast Safe to operate Versatile Accommodates Most Test Vials Select the Dual Block Model for Simultaneous Digestions
Easy-to-use and fast
The Hach DRB 200 Dry Thermostat Reactor provides a unique one-key operation. Programs for Hach procedures with digestion are preprogrammed into the instrument. And it’s fast—the block heats from 20 to 150°C in less than 10 minutes.
Safe to operate
The fully insulated heater block of the DRB 200 reactor means there can be no skin contact with the heater block. Temperature safeguards are provided to prevent overheating. The lids are transparent and the lock does deter premature checking of the progress of the reaction. And the reactor will emit an audible signal and automatically shut down at the end of the run.
Versatile
Use the DRB 200 reactor for digestions for metals analysis, digestions for nutrients analysis, or culture biological samples. Control temperatures in the reactor from 37 to 165°C in 1°C increments. In addition to preprogrammed digestion methods, use the reactor to program and store up to three custom methods.
Accommodates Most Test Vials
The DRB 200 reactor can heat solutions in round vials of two different sizes. Small, 16-mm diameter vial wells are suitable for Hach COD, UniCell™, TOC, and Test ’N Tube vials. Larger, 20-mm diameter vial wells are intended for sample preparation reaction vessels using the Metals Prep Set.
Select the Dual Block Model for Simultaneous Digestions
Two heat blocks in the DRB200 Dual Block Models reactor give the operator-independent control of two temperatures and durations. Use this option to run two programs at the same time. A Single Block DRB Reactor cannot be upgraded to a Dual Block Model at a later time.
TRUSTED_SUSTAINABILITY
Specifications
Type Photometers
Size Dual block (2 x 15 wells)