Learn More
Hach Company DRB200 Digital Reactor Block
Supplier:  Hach Company DRB20008
Encompass
DRB200 Digital Reactor Block for TNTplus 12x13mm vial wells-8x20 mm vial wells-230 VAC12 vials x 13 mm 8 vials x 20 mm (dual block) 230 Vac
Catalog No. 50-002-07603