Learn More
Hach Company DRB200 Digital Reactor Block
Supplier:  Hach Company DRB20006
Encompass
DRB200 Digital Reactor Block for TNTplus 21x13mm vial wells-4x20mm vial wells-230 VAC21 vials x 13 mm 4 vials x 20 mm (dual block) 230 Vac
Catalog No. 50-002-07783