Learn More
NEOGEN DRBC Agar (ISO), 5 kg
SDP
Catalog No. 502015394
Encompass_Preferred
Dichloran Rose Bengal Chloramphenicol Agar is for the enumeration of yeasts and is not intended for use in the diagnosis of disease or other conditions in humans.Developed with reference to ISO 21527-1:2008 for the enumeration of yeasts and molds in food and animal products, this medium is tested to the performance requirements of the standard. Used for the enumeration of viable yeasts and molds in products with a water activity of greater than 0.95 such as eggs, meat, some dairy products, fresh pastes, fruit and vegetables, DRBC Agar is designed to suppress the colonial growth of ‘spreader’ molds and in doing so allow easier performance of the colony count technique on yeasts and molds.For more information visit https://www.neogen.com/solutions/Microbiology/drbc-agar-iso
Catalog No. 50-201-5394 Supplier NEOGEN Supplier No. 700004471
Dichloran Rose Bengal Chloramphenicol Agar is for the enumeration of yeasts and is not intended for use in the diagnosis of disease or other conditions in humans.Developed with reference to ISO 21527-1:2008 for the enumeration of yeasts and molds in food and animal products, this medium is tested to the performance requirements of the standard. Used for the enumeration of viable yeasts and molds in products with a water activity of greater than 0.95 such as eggs, meat, some dairy products, fresh pastes, fruit and vegetables, DRBC Agar is designed to suppress the colonial growth of ‘spreader’ molds and in doing so allow easier performance of the colony count technique on yeasts and molds.For more information visit https://www.neogen.com/solutions/Microbiology/drbc-agar-iso