Learn More
TRIBIOSCIENCE INC Direct FFPE Tissue DNA Magnetic Extraction Kit, 200 Assays
SDP
Catalog No. NC2887505
Encompass_Preferred
Direct FFPE Tissue DNA Extraction Kit is designed for the purification of high-quality genomic DNA from formalin-fixed, paraffin-embedded (FFPE) tiny tissue samples. With a simple process, optimized lysis condition, and high-performance magnetic particles, this kit is capable of isolating high-quality DNA at most liquid handling workstations in the market.
Catalog No. NC2887505 Supplier TRIBIOSCIENCE INC Supplier No. TBS6028
Direct FFPE Tissue DNA Extraction Kit is designed for the purification of high-quality genomic DNA from formalin-fixed, paraffin-embedded (FFPE) tiny tissue samples. With a simple process, optimized lysis condition, and high-performance magnetic particles, this kit is capable of isolating high-quality DNA at most liquid handling workstations in the market.