Learn More
Eurofins Discoverx ChemiScreen D2 Dopamine Receptor Membrane Preparation, 1 vial. (HTS039M)
Catalog No. 502005727
Encompass
The ChemiScreen D2 Dopamine Receptor Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of DRD2 surface expression. ChemiScreen membrane preps can be used to screen for GPCR activity using GTPγS assays. Increased GPCR activation levels will lead to higher radioactive signal. Membrane preps contain 1 mL of 1 mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression. For detailed product information, visit www.discoverx.com/membranepreps.
Catalog No. 50-200-5727 Supplier Eurofins Discoverx Supplier No. HTS039M
The ChemiScreen D2 Dopamine Receptor Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of DRD2 surface expression. ChemiScreen membrane preps can be used to screen for GPCR activity using GTPγS assays. Increased GPCR activation levels will lead to higher radioactive signal. Membrane preps contain 1 mL of 1 mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression. For detailed product information, visit www.discoverx.com/membranepreps.