Learn More
Eurofins Discoverx Ready-to-Assay D4 Dopamine Receptor Frozen Cells, 2 Vials of Cells. (HTS223RTA)
Catalog No. 502006072
Encompass
The Ready-to-Assay D4 Dopamine Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. dopamine) based activation of DRD4 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses DRD4 alongside exogenous levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of DRD4, the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the stably modified clonal cell line in a convenient thaw-and-use format. For detailed product information, User manual and datasheet, visit www.discoverx.com/products-applications/frozen-cells.
Catalog No. 50-200-6072 Supplier Eurofins Discoverx Supplier No. HTS223RTA
The Ready-to-Assay D4 Dopamine Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. dopamine) based activation of DRD4 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses DRD4 alongside exogenous levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of DRD4, the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the stably modified clonal cell line in a convenient thaw-and-use format. For detailed product information, User manual and datasheet, visit www.discoverx.com/products-applications/frozen-cells.