Learn More
Medchemexpress LLC Dreadd agonist 21 | 56296-18-5 | 99.5% | 278.35 | C17H18N4 | 5 MG
SDP
Catalog No. 5000215617
Encompass_Preferred
DREADD agonist 21 is a potent chemogenetic ligand that selectively activates muscarinic DREADDs, exhibiting an EC50 of 1.7 nM at hM3Dq. Supplied as a light yellow solid, it is intended for research use in vitro and in vivo to modulate neuronal activity.
  • High potency at hM3Dq with EC50 1.7 nM.
  • High chemical purity (≈99.5%).
  • Available in small research quantities including 5 MG.
  • Stable solid form, light yellow to yellow appearance.
  • Recommended storage: powder -20°C for long-term stability.
  • Suitable for in vitro and in vivo chemogenetic studies.
Catalog No. 50-002-15617 Supplier Medchemexpress LLC Supplier No. HY1002345MG
DREADD agonist 21 is a potent chemogenetic ligand that selectively activates muscarinic DREADDs, exhibiting an EC50 of 1.7 nM at hM3Dq. Supplied as a light yellow solid, it is intended for research use in vitro and in vivo to modulate neuronal activity.
  • High potency at hM3Dq with EC50 1.7 nM.
  • High chemical purity (≈99.5%).
  • Available in small research quantities including 5 MG.
  • Stable solid form, light yellow to yellow appearance.
  • Recommended storage: powder -20°C for long-term stability.
  • Suitable for in vitro and in vivo chemogenetic studies.