Learn More
Medchemexpress LLC DREADD agonist 21 dihydrochloride | 2250025-92-2 | 98.9% | 351.27 g/mol | C17H20Cl2N4 | 1 ML
SDP
Catalog No. 5000215532
Encompass_Preferred
DREADD agonist 21 dihydrochloride is a water-soluble dihydrochloride salt of a potent chemogenetic ligand that selectively activates designer muscarinic receptor hM3Dq (EC50 = 1.7 nM). It is supplied for research use and is used to remotely control neuronal activity in vitro and in vivo studies.
  • Potent, selective hM3Dq agonist (EC50 = 1.7 nM).
  • Water-soluble dihydrochloride salt for improved handling and formulation.
  • Available as a solid and as a 10 mM solution in DMSO for flexible use.
  • High purity (~98.9%) suitable for research applications.
  • High solubility in DMSO and water to facilitate stock preparation.
Catalog No. 50-002-15532 Supplier Medchemexpress LLC Supplier No. HY100234A10MM 1ML
DREADD agonist 21 dihydrochloride is a water-soluble dihydrochloride salt of a potent chemogenetic ligand that selectively activates designer muscarinic receptor hM3Dq (EC50 = 1.7 nM). It is supplied for research use and is used to remotely control neuronal activity in vitro and in vivo studies.
  • Potent, selective hM3Dq agonist (EC50 = 1.7 nM).
  • Water-soluble dihydrochloride salt for improved handling and formulation.
  • Available as a solid and as a 10 mM solution in DMSO for flexible use.
  • High purity (~98.9%) suitable for research applications.
  • High solubility in DMSO and water to facilitate stock preparation.