Learn More
3D Universe Enclosed FFF 3D printer, 255 x 155 x 170 mm build volume, All-metal 0.4 mm hot end, Heated tempered glass bed to 100°C, 1.75 mm filament, 4.5 in color touchscreen, Wi-Fi/Ethernet/USB
SDP
Catalog No. NC1830314
Encompass
A professional-grade FFF desktop 3D printer designed for high-precision, reliable additive manufacturing in scientific, educational, and prototyping environments.
  • Fully enclosed, temperature-controlled build chamber for improved material compatibility and print consistency.
  • Single direct-drive extruder with an all-metal hot end and 0.4 mm nozzle for fine detail and engineering materials.
  • Max extruder temperature 280°C to support high-temperature filaments.
  • Heated tempered glass build plate with max bed temperature 100°C and build volume 255 x 155 x 170 mm.
  • Layer resolution options from 50 to 340 micron for a range of surface-finish needs.
  • 4.5 in diagonal full-color IPS touchscreen for local control and 8 GB internal storage for job management.
  • Connectivity via USB, Wi-Fi, and Ethernet with support for STL, OBJ, and gcode file formats.
  • Catalog No. NC1830314 Supplier 3D Universe Supplier No. 3D4501
    A professional-grade FFF desktop 3D printer designed for high-precision, reliable additive manufacturing in scientific, educational, and prototyping environments.
  • Fully enclosed, temperature-controlled build chamber for improved material compatibility and print consistency.
  • Single direct-drive extruder with an all-metal hot end and 0.4 mm nozzle for fine detail and engineering materials.
  • Max extruder temperature 280°C to support high-temperature filaments.
  • Heated tempered glass build plate with max bed temperature 100°C and build volume 255 x 155 x 170 mm.
  • Layer resolution options from 50 to 340 micron for a range of surface-finish needs.
  • 4.5 in diagonal full-color IPS touchscreen for local control and 8 GB internal storage for job management.
  • Connectivity via USB, Wi-Fi, and Ethernet with support for STL, OBJ, and gcode file formats.
  • Product Title