Learn More
Grainger Haws Axion MSR Eye/Face Wash, Deck Mounted, 3.7 GPM, 8 ft Hose, 1 ea
Catalog No. 502791428
Encompass_Preferred
The deck-mounted drench hose unit provides efficient emergency eye and face irrigation, utilizing a design that saves space while maintaining high performance. It features a specialized spray head that directs fluid to sweep contaminants away from the nasal cavity, ensuring effective decontamination in laboratory and industrial environments. This system is designed for quick activation during an emergency, supporting workplace safety compliance.
  • Features an AXION® MSR eye/face wash head with inverted directional laminar flow to achieve zero vertical velocity.
  • Includes a 2.44 m (8 ft.) swivel hose rated at 17.2 bar (250 psi) for extended reach and durability.
  • Utilizes a chrome-plated brass, stay-open squeeze lever valve for simple, continuous flow operation.
  • Equipped with a Noryl plastic deck flange featuring an integral key slot for secure positioning.
  • Includes a universal emergency sign
Catalog No. 50-279-1428 Supplier Grainger Supplier No. 16D384
The deck-mounted drench hose unit provides efficient emergency eye and face irrigation, utilizing a design that saves space while maintaining high performance. It features a specialized spray head that directs fluid to sweep contaminants away from the nasal cavity, ensuring effective decontamination in laboratory and industrial environments. This system is designed for quick activation during an emergency, supporting workplace safety compliance.
  • Features an AXION® MSR eye/face wash head with inverted directional laminar flow to achieve zero vertical velocity.
  • Includes a 2.44 m (8 ft.) swivel hose rated at 17.2 bar (250 psi) for extended reach and durability.
  • Utilizes a chrome-plated brass, stay-open squeeze lever valve for simple, continuous flow operation.
  • Equipped with a Noryl plastic deck flange featuring an integral key slot for secure positioning.
  • Includes a universal emergency sign