Learn More
Uline Drench Hose Eyewash Station - Deck Mount | | 1 EA
Catalog No. NC2738637
Encompass
Use as an eyewash station or pull drench hose to rinse your body, Twin spray heads with flip-top dust covers, Built-in filters for clean water, 8 PVC hose, Squeeze-handle with locking clip for hands-free use, Plumbing required, Recommended for labs and warehouses, Base flange allows for countertop installation, Includes eyewash station emergency sign and inspection tag, Meets ANSI Z3581-2014
Catalog No. NC2738637 Supplier Uline Supplier No. H9459
Use as an eyewash station or pull drench hose to rinse your body, Twin spray heads with flip-top dust covers, Built-in filters for clean water, 8 PVC hose, Squeeze-handle with locking clip for hands-free use, Plumbing required, Recommended for labs and warehouses, Base flange allows for countertop installation, Includes eyewash station emergency sign and inspection tag, Meets ANSI Z3581-2014