Learn More
MSC Bradley Drench Shower Tester, Nylon, 6 ft Handle, 1 ea
Catalog No. 501909651
Encompass
The Drench Shower Tester is a heavy-duty device designed to provide a convenient method for complying with ANSI specifications for weekly testing of all drench showers without the user getting wet. It features a watertight, chemical-resistant nylon funnel that directs water to a drain or bucket (not included), thereby preventing splashing in the surrounding area.
  • Watertight, chemical-resistant nylon funnel
  • Easy-to-reach, 6 ft aluminum handle
  • Prevents splashing in the surrounding area
  • Facilitates compliance with ANSI specifications
  • One-year warranty
Catalog No. 50-190-9651 Supplier MSC Supplier No. 01953637
The Drench Shower Tester is a heavy-duty device designed to provide a convenient method for complying with ANSI specifications for weekly testing of all drench showers without the user getting wet. It features a watertight, chemical-resistant nylon funnel that directs water to a drain or bucket (not included), thereby preventing splashing in the surrounding area.
  • Watertight, chemical-resistant nylon funnel
  • Easy-to-reach, 6 ft aluminum handle
  • Prevents splashing in the surrounding area
  • Facilitates compliance with ANSI specifications
  • One-year warranty