Learn More
Med Vet International Jorvet 200ml Drencher, J0641
Catalog No. 5000495933
Encompass_Preferred
Jorvet's 200ml drencher allows for administration of large doses often necessary to treat adult cattle. The use of oral calcium or propylene glycol drenches are easier with large drencher. Curved nozzle with olive tip Clear barrel to see medication High impact plastic handle
Catalog No. 50-004-95933 Supplier Med Vet International Supplier No. JOR641
Jorvet's 200ml drencher allows for administration of large doses often necessary to treat adult cattle. The use of oral calcium or propylene glycol drenches are easier with large drencher. Curved nozzle with olive tip Clear barrel to see medication High impact plastic handle