Learn More
Med Vet International Jorvet 300ml Drencher, J0640
Catalog No. 5000495875
Encompass_Preferred
Jorvet's 300ml drencher allows for administration of large doses often necessary to treat adult cattle. The use of oral calcium or propylene glycol drenches are easier with large drencher. Curved nozzle with olive tip Clear barrel to see medication High-impact plastic handle
Catalog No. 50-004-95875 Supplier Med Vet International Supplier No. JOR640
Jorvet's 300ml drencher allows for administration of large doses often necessary to treat adult cattle. The use of oral calcium or propylene glycol drenches are easier with large drencher. Curved nozzle with olive tip Clear barrel to see medication High-impact plastic handle