Learn More
Med Vet International Speedy Calf Drencher, 4 Liter, J1317L, Each
Catalog No. 5000495951
Encompass_Preferred
The premium oral calf feeder. - Allows a one-handed delivery - Multi-directional easy-grip - Three speed switch to control air flow thus fluid outflow - Soft rubber probe tip with lateral eyes for less trauma and likelihood of blockage - Large 3" (70mm) opening for easy filling and cleaning - Bottle stands up on flat surface - Probe bends at bottle via rubber connection so flow starts when desired Speedy Calf Drencher 4.0L
Catalog No. 50-004-95951 Supplier Med Vet International Supplier No. JOR1317L
The premium oral calf feeder. - Allows a one-handed delivery - Multi-directional easy-grip - Three speed switch to control air flow thus fluid outflow - Soft rubber probe tip with lateral eyes for less trauma and likelihood of blockage - Large 3" (70mm) opening for easy filling and cleaning - Bottle stands up on flat surface - Probe bends at bottle via rubber connection so flow starts when desired Speedy Calf Drencher 4.0L