Learn More
Med Vet International Foam Dressing Allevyn Life 9-4/5 X 9-9/10 Inch With Border Film Backing Silicone Gel Adhesive Heel Sterile, 66801304, 30/Case
Supplier: Med Vet International 863007CS
Multi-layered design incorporates hydrocellular foam, a hyper-absorber lock-away core and a masking layer that has been designed for comfort Design provides coverage to the malleolus without the need for secondary retention Masking layer minimizes the visual impact of strike-through, providing the wearer with confidence that her dressing may not attract the negative attention of others Excellent fluid locking under pressure Silicone wound contact layer minimizes trauma to the wound and pain during dressing changes Heel dressing is 25 x 25.2 centimeters