Learn More
Med Vet International Oil Emulsion Impregnated Dressing 3M Adaptic Rectangle 3 X 8 inch Sterile, 2013, 3/Pack, 36 Packs
Catalog No. 502829068
Encompass_Preferred
Knitted cellulose acetate is impregnated with petrolatum emulsion to allow exudate to easily pass through to the absorbent dressing Nonadherent to prevent dressing from sticking to wounds Can be cut or trimmed without unraveling or shredding
Catalog No. 50-282-9068 Supplier Med Vet International Supplier No. 684062BX
Knitted cellulose acetate is impregnated with petrolatum emulsion to allow exudate to easily pass through to the absorbent dressing Nonadherent to prevent dressing from sticking to wounds Can be cut or trimmed without unraveling or shredding