Learn More
Med Vet International Adhesive Dressing 3M Medipore 3-1/2 X 4 inch Soft Cloth Rectangle White Sterile, 3566, 100/Case
Catalog No. 502831733
Encompass_Preferred
3M's Medipore +Pad soft cloth dressings are porous and breathable to maintain skin integrity They have a nonadherent pad that won't unsettle the wound bed 1-3/4 inch x 2-3/8 inch pads are absorbent to help maintain a proper healing environment The adhesive on the white, 3½ x 4 inch dressing is strong yet gentle on the skin These Medipore island dressings conform around contours and flex with movement to provide optimal protection
Catalog No. 50-283-1733 Supplier Med Vet International Supplier No. 315360CS
3M's Medipore +Pad soft cloth dressings are porous and breathable to maintain skin integrity They have a nonadherent pad that won't unsettle the wound bed 1-3/4 inch x 2-3/8 inch pads are absorbent to help maintain a proper healing environment The adhesive on the white, 3½ x 4 inch dressing is strong yet gentle on the skin These Medipore island dressings conform around contours and flex with movement to provide optimal protection