Learn More
Med Vet International Dynarex Sterile Dynaginate Calcium Alginate Dressing, 4" X 8", 60/Case
Catalog No. 502831331
Encompass_Preferred
Calcium Alginate Dressings contours to wound bed and changes to a fibrous gel when in contact with wound exudate, creating an optimal moist wound environment important to healing. Changes to a fibrous gel when in contact with wound exudate, creating a moist wound environment. Contours to the wound bed and will not adhere to the wound upon removal. High absorbency dressing used for heavily exuding wounds. Primary dressing ideal for the management of partial and full thickness wounds at stages 3 or 4.
Catalog No. 50-283-1331 Supplier Med Vet International Supplier No. 3028
Calcium Alginate Dressings contours to wound bed and changes to a fibrous gel when in contact with wound exudate, creating an optimal moist wound environment important to healing. Changes to a fibrous gel when in contact with wound exudate, creating a moist wound environment. Contours to the wound bed and will not adhere to the wound upon removal. High absorbency dressing used for heavily exuding wounds. Primary dressing ideal for the management of partial and full thickness wounds at stages 3 or 4.